Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint, with PUR Coating
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03044037
 View more versions of this product
Product Code. 11903391
  • 92.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Especially for rotary evaporators
  • With center neck and conical joint
  • With highly transparent PUR coating
  • Autoclavable
  • Pear-shaped
  • Material: DURAN™
Specifications
Specifications
Duran Glass
100 mL
60 mm