Learn More
Evocalcet, MedChemExpress
MedChemExpress Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Brand: Medchem Express HY-17613
This item is not returnable.
View return policy
Description
Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.Specifications
Evocalcet | |
Off-White | |
C24H26N2O2 | |
KHK7580 | |
O=C(O)CC1=CC=C(N2C[C@@H](N[C@@H](C3=C4C=CC=CC4=CC=C3)C)CC2)C=C1 | |
374.48 | |
Research | |
Metabolism-protein/nucleotide metabolism |
870964-67-3 | |
Research | |
50 mg | |
DMSO : 30 mg/mL (80.11 mM; Need ultrasonic and warming) | |
374.48 | |
99.05% | |
Solid |