Learn More
Evocalcet, MedChemExpress
MedChemExpress Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Brand:  Medchem Express HY-17613
Encompass_Preferred
 View more versions of this product
Product Code. 16479897
  • 117.45 € / 5mg
Explore more special offers
Description
Description
Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Specifications
Specifications
Evocalcet
Off-White
C24H26N2O2
KHK7580
O=C(O)CC1=CC=C(N2C[C@@H](N[C@@H](C3=C4C=CC=CC4=CC=C3)C)CC2)C=C1
374.48
Research
Metabolism-protein/nucleotide metabolism
870964-67-3
Research
5 mg
DMSO : 30 mg/mL (80.11 mM; Need ultrasonic and warming)
374.48
99.05%
Solid