Learn More
Thermo Scientific™ eVol™ XR Dispensing System Syringes
Rely on these versatile and functional syringes in sizes from 5 to 1000μL for the Thermo Scientific™ eVol™ Dispensing System.
124.00 € - 146.36 €
Specifications
For Use With (Equipment) Electronic Dispensing System
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10558995
View Documents
Thermo Scientific™
66002-021
5 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10488944
View Documents
Thermo Scientific™
66002-022
50 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10302725
View Documents
Thermo Scientific™
66002-023
500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15230369
View Documents
Thermo Scientific™
66002-038
100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Rely on exceptional versatility and functionality when using Thermo Scientific™ eVol™ XCHANGE Syringes. For use with the unique, hand-held eVol XR Dispensing System, the syringes are available in sizes from 5 to 1000μL. Integrated XCHANGE™ coupling allows you to change syringes quickly and easily. The addition of MEPS to XCHANGE syringe offers automated sample preparation by SPE in a handheld system. The eVol XR Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures
The eVol XR Dispensing System is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol system is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibly for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Specifications
Specifications
Electronic Dispensing System
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ eVol™ XR Dispensing System Syringes