missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Evolution™ 260 Bio UV-Visible Spectrophotometer
Shop All Thermo Scientific Products
Click to view available options
Type:
Evolution 260 Bio Computer Ctl
Includes:
EU/US/UK power cords
Unit Size:
Each
Description
New sales of this model have been discontinued. Please see the newest models: the Evolution Pro Spectrophotometers.
Complete Life Science Applications
Next generation INSIGHT software provides the streamlined convenience of easy-to-understand nucleic acid and protein analysis methods and includes dedicated modules for:
- Nucleic Acid concentration by direct UV measurement at 260nm including DNA, ssDNA, RNA, and siRNA
- Nucleic acid purity measurements from 260/280nm and 260/230 ratios
- Protein analysis by direct UV measurement at 205 or 280nm
- Most common colorimetric analysis methods including: Pierce 660nm Protein assay, Bradford, BCA and Lowry
- Measuring the fluorescence labeling efficiency for proteins and nucleic acids
- Measuring the microarray labeling efficiency with one or two dyes
- Customizable, Straightforward INSIGHT Software
Personalize your home screen for easy navigation and convenience
- Workbook tools save time and ensure accurate results
- CUE scripting allows creation of customized interface and simplified workflow
Best-in-Class Performance
- Double-beam geometry ideal for kinetics or any sample that may change over time
- Application Focused Beam Geometry (AFBG) technology provides superior results for microcells and turbid samples
- Xenon flash lamp requires no warm-up time allowing instant measurements
- Electronic triggering provides the highest level of accuracy for rapid-mixing kinetics measurements
- Precision monochromator drive delivers fast scanning data collection and wavelength accuracy Powerful Convenience at your Fingertips
- Color, touch screen provides instrument control using built-in computer
- Integrated keypad allows communication with INSIGHT software
- Choice of Local or PC configurations provides flexibility and convenience
Accessories for all your Sampling Needs
- Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
- Automated cell changers with Peltier temperature control increase sample throughput
- Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100°C
- Easy software integration gives you complete control of your measurements
Reliable Assurance of System Performance
- The Thermo Scientific UV Validator package provides support for system qualification and validation activities
- Complete documentation facilitates compliance of your system with the requirements of FDA, GxP, ISO 9001:2000 and ISPE 2001 guidelines and regulations
- CVC and Mercury Lamp accessory automated testing ensures accuracy of your data
- Thermo Scientific INSIGHTsecurity software provides complete tools to meet 21 CFR Part 11 requirements
Specifications
Specifications
| Baseline Flatness | ±0.0010 A, 200 to 800 nm, 1.0 nm SBW, smoothing |
| Certifications/Compliance | ISO 9001:2008 |
| Detector Type | Dual Silicon Photodiodes |
| Dimensions (L x W x H) | 62.2 x 48.6 x 27.9 cm (24 x 19 x 11 in.) |
| Product Line | Evolution 260 Bio |
| Spectral Bandwidth | Variable: 1.0 nm, 2.0 nm |
| Type | Evolution 260 Bio Computer Ctl |
| Wavelength Range | 190 to 1100 nm |
| Drift | <0.0005 A/hr., 500 nm, 1.0 nm SBW, 1 hr. warmup |
| Electrical Requirements | 100/240 V, 50/60 Hz selected automatically, 150 W maximum |
| Show More |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu