missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometers

Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.

9755.00 € - 26780.00 €


Products 6
Product Code Brand Description Spectral Bandwidth Price Quantity & Availability  
Product Code Brand Description Spectral Bandwidth Price Quantity & Availability  
17329263
View Documents
Thermo Scientific™
840-341400
Evolution One UV-Vis Spectrophotometer 1 nm
9755.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17339263
View Documents
Thermo Scientific™
840-341500
Evolution One Plus UV-Vis Spectrophotometer 1.0, 2.0 nm Variable
11235.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17435603
View Documents
Thermo Scientific™
840-370500

Evolution One Security and Validator Bundle includes:
-Evolution One UV-Vis Spectrometer
-INSIGHT Pro with Security Software
-Validator for Insight Pro A4 Paper Version
-Qualification Filter Kit for UV-Vis

1 nm
14230.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17445603
View Documents
Thermo Scientific™
840-370600
Evolution One USP/EP Standards Pharma Bundle includes:
-Evolution One UV-Vis Spectrometer
-INSIGHT Pro with Security Software
-Validator Documentation A4 Paper Version
-USP and EP UV Standards Set
-USP and EP Visible Standards Set
1 nm
24145.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17465603
View Documents
Thermo Scientific™
840-370800
Evolution One Plus Security and Validator Bundle includes:
-Evolution One Plus UV-Vis Spectrometer
-INSIGHT Pro with Security Software
-Validator for Insight Pro A4 Ltr Paper Version
-Qualification Filter Kit for UV-Vis
1.0, 2.0 nm Variable
16840.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17455603
View Documents
Thermo Scientific™
840-370700
Evolution One Plus USP/EP Standards Pharma Bundle includes:
-Evolution One Plus UV-Vis Spectrometer
-INSIGHT Pro with Security Software
-Validator Documentation A4 Ltr Paper Version
-USP and EP UV Standards Set
-USP and EP Visible Standards Set
1.0, 2.0 nm Variable
26780.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For routine, complex research and advanced applications, including current compliance, the Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometer series brings together reliable, versatile hardware, a broad range of accessories, with easy-to-use, easy-to-learn Thermo Scientific™ Insight™ Pro Software.

Webinar: Identification of protein aggregation through UV-Vis spectroscopy

For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.

Optimized Engineering Delivers Ultimate Performance:
  • Double-beam geometry with fixed 1nm or variable spectral bandwidth options
  • Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories
Insight Pro Software—Easy-to-learn, easy-to-use
  • Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
  • Prevent costly sampling and measurement errors: Create dedicated workflows for complex assays and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
Accessories for all your Sampling Needs:
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
  • Easy software integration gives you complete control of your measurements

Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Evolution One/One Plus Security and Validator Bundles include:
  • Evolution One or One Plus UV-Vis Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • Qualification Filter Kit for UV-Vis
Evolution One/One Plus USP/EP Standards Pharma Bundles include:
  • Evolution One or One Plus Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • USP and EP UV Standards Set
  • USP and EP Visible Standards Set
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.