missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ExcelGel 12.5%, Tris-Acetate, 0.5 mm, Protein Gel, 25-well

Separation Range: 15 to 40 kd; Separation Type: Denaturing; For Use With: Immobiline DryStrip Gels

Brand:  Cytiva 80126101

833.26 EUR valid until 2024-12-31
Use promo code "21959" to get your promotional price.


Product Code. 11753339

  • 934.00 € / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Three homogeneous and one gradient polyacrylamide gel for horizontal electrophoresis of SDS denatured proteins; also for separation of nucleic acids in the base pair range 50-1500

  • Homogeneous 7.5, 12.5, and 15% gels have 25 preformed sample wells for sample volumes from up to 17μL for quick, easy sample application
  • Gradient gel uses sample application strips (26 samples up to 40μL, 52 samples up to 20μL), paper pieces, or sample droplets; recommended for use with Immobiline DryStrip gels
  • ExcelGel SDS Homogeneous 15% is also designed for separation of peptides

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyester
Immobiline DryStrip Gels
0.5 mm
Homogeneous Page
15 to 40 kDa
25-well
Polyester film
6% T/3% C
0.019 in.
4.3 in.
11 cm
9.8 in.
25 cm
ExcelGel precast gels for SDS-PAGE
12.5%
Tris-Acetate
Protein
Denaturing
Analytical Chemistry and Research, Research Horizontal Electrophoresis of SDS Denatured Proteins
6/Pk.
2 - 8°C
4.3 in.
110 mm
9.8 in.
25 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-90 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 90% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers