missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ExcelGel 8 to 18%, Tris-Acetate, 0.5 mm, Protein Gel, 25-well

Separation Range: 15 to 200 kd; For Use With: Immobiline DryStrip Gels

Brand:  Cytiva 80125553

Product Code. 11733339

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Three homogeneous and one gradient polyacrylamide gel for horizontal electrophoresis of SDS denatured proteins; also for separation of nucleic acids in the base pair range 50-1500

  • Homogeneous 7.5, 12.5, and 15% gels have 25 preformed sample wells for sample volumes from up to 17μL for quick, easy sample application
  • Gradient gel uses sample application strips (26 samples up to 40μL, 52 samples up to 20μL), paper pieces, or sample droplets; recommended for use with Immobiline DryStrip gels
  • ExcelGel SDS Homogeneous 15% is also designed for separation of peptides

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyester
Immobiline DryStrip Gels
0.5 mm
Gradient Page
15 to 200 kDa
Analytical Chemistry and Research, Research Horizontal Electrophoresis of SDS Denatured Proteins
6/Pk.
20 - 30°C
4.3 in.
110 mm
9.6 in.
24.5 cm
ExcelGel precast gels for SDS-PAGE
8 to 18%
Tris-Acetate
Protein
25-well
Polyester film
5% T/3% C
0.019 in.
4.3 in.
11 cm
9.6 in.
24.5 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-90 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 90% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers