Learn More
R&D Systems™ ExCellerate Human T Cell Expansion Media, Xeno-Free
Media for the ex vivo expansion and culture of human T cell lymphocytes under xeno-free conditions
Brand:  R&D Systems™ CCM030
101.94 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18777146
  • 130.00 € / 500mL
Explore more special offers
Description
Description
  • Robust T cell expansion under serum-free and xeno-free conditions
  • Optimized for use with the Cloudz™ T Cell Activation Kit
  • Compatible with a variety of T cell activation methods
  • Supports T cell expansion from PBMCs or purified CD3+ T cells
Specifications
Specifications
Store the unopened product at 2 - 8°C. Do not use past expiration date.
Facilitates discovery and preclinical research for T cell immunoregulation and therapy, robust platforms for ex vivo expansion and maintenance of T cells, including specialized serum-free or xeno-free media
T-Cell Expansion
500 mL