Learn More
R&D Systems™ ExCellerate iPSC Expansion Medium
Brand:  R&D Systems™ CCM036
Product Code. 18759296
  • 289.00 € / 500mL
Explore more special offers
Specifications
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
500 mL