Learn More
Eppendorf™ Exchangeable Thermoblock
Accessories for ThermoStat plus/thermoMixer plus
655.35 €
Specifications
No. of Places 24
Type Block
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10344651
View Documents Promotion Details
Eppendorf™
5361000015
24 Reaction Vessels 0.5mL with Work Rack and Cool Pack 0°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10439472
View Documents Promotion Details
Eppendorf™
5360000011
24 Reaction Vessels 1.5mL with Work Rack and Cool Pack 0°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Compact, bench top thermostat with thermoblocks for 0.2mL, 0.5mL, 1.0mL, 2.0mL tubes and microtitre plates designed to provide a high degree of flexibility for a multitude of applications
  • Programmable temperature and run time for special applications
  • GLP compliant
  • RS232 interface
Specifications
Specifications
24
Block