Learn More
asecos™ Exhaust Air Unit
Extraction module for wall mounting, tall cabinets, or under bench cabinets with exhaust air monitoring
Brand:  asecos™ 24732
 View more versions of this product
Product Code. 15957826
  • 1175.00 € / Each
Explore more special offers
Description
Description
  • Extraction units/modules for installation on the cabinet or wall mounting – ready to plug-in, easy and quick installation
  • Safe capturing of escaping vapours and gases at the point where they are released or produced
  • Very low noise, ideal for direct installation at the workplace
  • With connection cable and power cord
  • Connecting fitting DN 75
  • ATEX compliant: C` II 3/-G Ex c llC T4
  • With extraction air monitoring
    • Integrated pressure leak detector
    • The green LED turns off in case of failure and the red LED lights up
Specifications
Specifications
CE compliant
For wall mounting or under bench cabinets, with exhaust air monitoring and with potential-free alarm contact, CE compliant: C ` II 3/- G Ex c IIC T4, explosion-proof, extraction from: EX-Zone 2 / installation: outside any EX-ZONE
Extraction module for wall mounting – ready to plug-in, easy and quick installation
340 mm
HF.EA.24139
12 kg
Steel
200 mm
200 x 400 x 340 mm
Under bench cabinets
Installation bracket, flexible hose, hose clips and installation material
Powder-coated plain
400 mm
Exhaust air unit