missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exosome Standards (A549 cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
417.00 € - 1345.00 €
Specifications
Content And Storage | Store at 4°C. After reconstitution store at -70°C. |
---|---|
Concentration | LYOPH |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18644630
|
Novus Biologicals™
NBP3-11645-180ug |
6 x 30 μg Vials |
1080.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18696070
|
Novus Biologicals™
NBP3-11645-60ug |
2 x 30 μg Vials |
417.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18628441
|
Novus Biologicals™
NBP3-11645-600ug |
6 x 100 μg Vials |
1345.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18637851
|
Novus Biologicals™
NBP3-11645-200ug |
2 x 100 μg Vials |
530.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Spezifikation
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Novus Biologicals™ Exosome Standards (A549 cell line)