missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exosome Standards (COLO1 cell line)
Assay Kit (Colorimetric)
Brand: Novus Biologicals™ NBP2-49845-600ug
1076.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH | |
6 x 100 μg Vials |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Exosome Standards (COLO1 cell line) > 6 x 100ug Vials
Spot an opportunity for improvement?Share a Content Correction