Learn More
Novus Biologicals™ Exosome Standards (HT29 cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand:  Novus Biologicals™ NBP3-11685-600ug
 View more versions of this product
Product Code. 18623800
  • 1345.00 € / Pack of 6
Explore more special offers
Specifications
Store at 4°C. After reconstitution store at -70°C.
LYOPH
6 x 100 μg Vials