Learn More
Novus Biologicals™ Exosome Standards (LnCAP cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand:  Novus Biologicals™ NBP3-11687-60ug
 View more versions of this product
Product Code. 18687640
  • 417.00 € / Pack of 2
Explore more special offers
Specifications
Store at 4°C. After reconstitution store at -70°C.
LYOPH
2 x 30 μg Vials