missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exosome Standards (SK-N-SH cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
417.00 € - 1345.00 €
Specifications
Content And Storage | Store at 4°C. After reconstitution store at -70°C. |
---|---|
Concentration | LYOPH |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18604430
|
Novus Biologicals™
NBP2-49852-180ug |
6 x 30 μg Vials |
1080.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18694850
|
Novus Biologicals™
NBP2-49852-60ug |
2 x 30 μg Vials |
417.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18634031
|
Novus Biologicals™
NBP2-49852-600ug |
6 x 100 μg Vials |
1345.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18675380
|
Novus Biologicals™
NBP2-49852-200ug |
2 x 100 μg Vials |
529.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Exosome Standards (SK-N-SH cell line)