Learn More
Novus Biologicals™ Exosome Standards (SK-N-SH cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand:  Novus Biologicals™ NBP2-49852-600ug
 View more versions of this product
Product Code. 18634031
  • 1345.00 € / Pack of 6
Explore more special offers
Specifications
Store at 4°C. After reconstitution store at -70°C.
LYOPH
6 x 100 μg Vials