missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXPH5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22199-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
EXPH5 | |
Polyclonal | |
Unconjugated | |
EXPH5 | |
exophilin 5, EXPH5, KIAA0624, SLAC2 B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23086 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NEV8 | |
EXPH5 | |
EXPH5 Fusion Protein Ag17449 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction