missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ Expression Medium
GREENER_CHOICE

Expi293™ Expression Medium

398.00 € - 6760.00 €

Specifications

Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type Expression Medium
Product Line Expi293™
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15490694
View Documents
Gibco™
A1435103
10 L
3525.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
15400704
View Documents
Gibco™
A1435104
20 L
6760.00 €
20 L
Please to purchase this item. Need a web account? Register with us today!
13469756
GREENER_CHOICE
View Documents
Gibco™
A1435101
1 L
398.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
13489756
GREENER_CHOICE
View Documents
Gibco™
A1435102
6 x 1 L
2025.00 €
6 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Expi293™ Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F™ cells in the Expi293™ Expression System for scalable transient protein expression.

  • Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
  • Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine™ transfection reagent
  • Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
  • Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10L in disposable bioreactors
  • Does not contain phenol red

    Expi293™ Expression Medium is formulated with GlutaMAX™-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293™ Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.
  • Specifications

    Specifications

    Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Liquid
    Expression Medium
    Expi293™
    Videos
    SDS
    Documents

    Documents

    Product Certifications

    For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.