missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiFectamine™ CHO Transfection Kit

The ExpiFectamine™ CHO Transfection Kit is a core component of the ExpiCHO™ Expression System Kit, designed for transient transfection of high-density cultures of Chinese hamster ovary (CHO) cells.

Brand:  Gibco™ A29130

9679.80 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15627878

  • 12410.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

The ExpiFectamine CHO Transfection Kit is a core component of the ExpiCHO Expression System Kit (Cat. No. A29133) and enables transient transfection of high-density cultures of Chinese hamster ovary (CHO) cells. The high-efficiency, cationic transfection reagent, coupled with an optimized transfection enhancer and culture feed, will help power the highest possible level of protein expression from ExpiCHO-S Cells (Cat. No. A29127) cultured in ExpiCHO Expression Medium (Cat. No. A2910001).

All components work together synergistically with expression yields of 1–3 g per L of transfected culture demonstrated for some antibody and non-antibody proteins.

Features of the ExpiFectamine CHO Transfection Kit include:

  • Enables robust and reproducible transfection results, giving you greater confidence in your results and ability to use CHO cells from research to discovery
  • Achieve >90% transfection efficiency of high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
  • Uses one-third to one-half less plasmid DNA than other transfection reagents
  • Scale growth and transfection in culture formats from <1 mL in deep well plates up to 5 L when using Nalgene angled-bottom shake flasks
  • Flexible, reliable culture and transfection protocols with multiple options that readily fit into current transient expression workflows

The ExpiFectamine CHO Transfection Kit is specifically designed to work with ExpiCHO-S Cells in the ExpiCHO Expression System Kit. The transfection kit supplies the cationic ExpiFectamine CHO Transfection Reagent, ExpiFectamine CHO Transfection Enhancer, and ExpiCHO Feed. ExpiCHO-S Cells and ExpiCHO Expression Medium are available separately.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ExpiCHO-S™, CHO-S, SFM Adapted
Kit includes:
• ExpiFectamine CHO Reagent, 32 mL
• ExpiFectamine CHO Enhancer, 60 mL
• ExpiCHO Feed, 3.3 L

Sufficient for transfection of 10 L of culture
Store in the dark at 2°C to 8°C.
Lipid-based Transfection
CHO Transfection Kit
Room Temperature
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Ovarian Cells
ExpiFectamine™
For 10 L of culture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.