missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiSf™ Expression System Starter Kit

Complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow

Brand:  Gibco™ A39111

620.16 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 17144709

  • 1216.00 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The ExpiSf Expression System is a complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow.
  • A chemically defined, yeastolate-free, animal origin-free medium for consistent cell growth and protein expression
  • Integrated components that work in concert to deliver three times higher protein yields compared to traditional insect expression platforms
  • Robust production of high-titer, high-quality P0 recombinant baculovirus in suspension culture format without the need for further virus amplification
  • A fast, simplified workflow that allows you to obtain protein in as little as 6–10 days; half the time of traditional protocols
  • Scalable expression that can be applied to cultures from 24-deep well plates to >3 L shake flasks
  • The components includes in the ExpiSf Expression System Starter Kit are: 2 vials of frozen ExpiSf9 Cells, 1 liter of ExpiSf CD Medium, ExpiSf Enhancer sufficient for 1 liter of culture, and 1 mL ExpiFectamine Sf Transfection Reagent. All components of the ExpiSf Expression System are also available for purchase separately.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 1000 mL bottle ExpiSf CD Medium, store in the dark at 2–8°C
• 4 x 1 mL ExpiSf Enhancer, 2–8°C
• 1 mL ExpiFectamine Sf Transfection Reagent, 2–8°C
• 2 x 1 vial ExpiSf9 Cells, store in vapor-phase liquid nitrogen

Sf9, SFM Adapted
ExpiSf™
Approved for shipment at Room Temperature or on Wet or Dry Ice
Baculovirus
Insect Cell Expression System Starter Kit
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.