missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiFectamine™ Sf Transfection Reagent

Product Code. 15869126
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 15869126

missing translation for 'mfr': Gibco™ A38915

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Gibco™ ExpiSf™ Transfection Reagant is a cationic lipid-based reagent for efficient DNA transfection of insect cells using fast, flexible protocols.

  • Delivers robust and reproducible transfection, giving you greater confidence in your results
  • Enables transfection of plasmid DNA in adherent and suspension Sf9 and Sf21 cell cultures for reliable recombinant baculovirus production
  • Facilitates efficient production of high-titer, high-quality P0 recombinant baculovirus in suspension culture format without the need for further virus amplification
  • Exhibits low toxicity and does not require medium change post-transfection
  • Is part of a simple, fast protocol with a short 5-minutes complexation time
  • Demonstrates linear scalability of transfections from culture volumes of 4mL to 500mL in suspension format

The ExpiFectamine Sf Transfection Reagent is a core component of the ExpiSf Expression System and has been optimized for transfection of ExpiSf9™ cells (Cat. No. A35243). However, the reagent also demonstrates superior performance using other Sf9 and Sf21 insect cells cultured in serum-free media such as Sf-900 II SFM or Sf-900 III SFM in both adherent and suspension culture formats.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C, do not freeze

Shelf Life: 24 months from date of manufacture

Cell Type Insect Cells
Sample Type Baculovirus DNA, Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility High-throughput Compatible
Product Line ExpiFectamine
Product Type Transfection Reagent
Quantity 1 mL
Serum Compatible Yes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.