Learn More
Tuttnauer Extend Cycle Time
Special program to work with extended sterilization times of more than 99 minutes. Must specify sterilization temperature (121°C or 134°C) and length of sterilization time.
Brand:  Tuttnauer ADD222-0458
Product Code. 17819538
  • 1680.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Special program to work with extended sterilization times of more than 99 minutes.
  • Must specify sterilization temperature (121°C or 134°C) and length of sterilization time.
Specifications
Specifications
Software
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer Extend Cycle Time > Extend Cycle Time