Learn More
Memmert™ Extended Overtemperature Protection for SN160
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ SN160A6
Product Code. 17508485
  • 195.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
SN160 Sterilizer
Integrated Pt100 Sensor