Learn More
Grant Instruments™ External Probe
TX external probes (optional) for monitoring and controlling temperature of remote loads.
Brand:  Grant Instruments™ TXPEP
 View more versions of this product
Product Code. 13163743
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Flexible plastic or Stainless steel probe
  • 3m cable (Din plug)
Specifications
Specifications
3m cable (DIN plug)
Plastic