missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ External Probe
For Use With: Isotemp (±1°C at 70°C) and Isotemp Advanced (±0.5°C at 70°C) Hotplates and Hotplate Stirrers
Brand: Fisherbrand™ CIC0001466
64.50 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Optimize performance and flexibility

Specifications
Isotemp (±1°C at 70°C) and Isotemp Advanced (±0.5°C at 70°C) Hotplates and Hotplate Stirrers | |
0°C to 400°C | |
Stainless Steel |
Stainless Steel | |
Probe |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ External Probe > Isotemp +/-1.0C at 70C
Spot an opportunity for improvement?Share a Content Correction