missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves
Click to view available options
Size:
10
11
6.5 to 11
7.5
8
9
Unit Size:
12 pairs
Pack of 144
Description
- 50% thicker than standard flocklined gloves
- Chlorinated for better grip and increased chemical resistance
- 100% natural rubber with no fillers for excellent resistance to mechanical hazards
- 100% cotton flocking for softness, comfort and increased absorption
- Ideal for chemical processing and preparation and maintenance of plant and heavy equipment
Specifications
Specifications
| Back Material | Rubber Latex |
| Certifications/Compliance | EN 374 |
| Cuff Material | Natural Rubber Latex |
| Coating Material | Natural Rubber Latex |
| Color | Orange |
| Latex Free | No |
| Product Type | Immersion Gloves |
| Size | 7.5 |
| Style | 87-955 |
| Thickness (Metric) | 0.75 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction