missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves
Provides for heavy duty handling where sensitivity is also required. Ansell Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves offer extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
36.20 € - 447.58 €
Specifications
Back Material | Rubber Latex |
---|---|
Cuff Material | Natural Rubber Latex |
Coating Material | Natural Rubber Latex |
Latex Free | No |
Product Type | Immersion Gloves |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12687041
|
Ansell™
87955070 |
7.5 | N/A | ||||||
11786564
|
Ansell™
87955080BP |
8 |
54.40 €
12 pairs |
In Stock
Log in to see stock. |
|||||
11776564
|
Ansell™
87955090BP |
9 |
54.40 €
12 pairs |
In Stock
Log in to see stock. |
|||||
11716564
|
Ansell™
87955100BP |
10 |
54.40 €
12 pairs |
In Stock
Log in to see stock. |
|||||
12677041
|
Ansell™
87955110 |
11 |
54.40 €
12 pairs |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15248003
|
Ansell™
87-955_6.5-7 |
6.5 to 11 | N/A | ||||||
Description
- 50% thicker than standard flocklined gloves
- Chlorinated for better grip and increased chemical resistance
- 100% natural rubber with no fillers for excellent resistance to mechanical hazards
- 100% cotton flocking for softness, comfort and increased absorption
- Ideal for chemical processing and preparation and maintenance of plant and heavy equipment
Specifications
Rubber Latex | |
Natural Rubber Latex | |
Immersion Gloves | |
0.75 mm | |
Straight | |
144Pack | |
87-955 |
Natural Rubber Latex | |
No | |
87-955 | |
Orange | |
Cotton Flocking | |
Many ketones, salts, detergents, alcohols, alkalis and fats |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title