Learn More
Velp Scientifica™ Extraction Thimble Holder
Velp Scientifica™ Extraction Thimble Holder for use with SER 148 Series Solvent Extractors.
Brand:  Velp Scientifica™ A00001142
Product Code. 10120464
  • 43.55 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with SER 148 Series Solvent Extractors
Specifications
Specifications
SER 148 Series Solvent Extractor
Extraction Thimble Holder
1 Ea.