missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Extraction Thimbles, Glass Fiber, MN 649

Provide short-term temperature resistance up to 500°C.

Brand:  Macherey Nagel 649122

Product Code. 10595102

  • 221.00 € / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Made from glass microfibers without binder
  • Short-term temperature resistance up to 500°C
  • Very high particle retention for dust analysis in hot exhaust gases
Specifications

Specifications

Borosilicate Glass Fiber
25/Pk.
94 mm
Short-term temperature resistance, high particle retention, for dust analysis in hot exhaust gases
33 mm ID in accordance with DIN 12449 for extractors with defined nominal volumes according to DIN 12602 and 12604
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers