missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Extraction Thimbles Porosity 0
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
56.00 € - 73.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10050241
|
Lenz Laborglasintrumente
05405243 |
150 mL |
58.00 €
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
10316361
|
Lenz Laborglasintrumente
05405225 |
30 mL |
56.00 €
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
10797891
|
Lenz Laborglasintrumente
05405249 |
250 mL |
73.00 €
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
Description
- Extraction thimbles
- Glass insert with bores for solids
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) | |
0 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title