Learn More
asecos™ Extraction Unit
Extraction units/modules for installation on the cabinet or wall mounting – ready to plug-in, easy and quick installation
Brand:  asecos™ 14218
Product Code. 10686644
  • 762.00 € / Each
Explore more special offers
Alert:
For indoor use only
Description
Description
Extraction units/modules for installation on the cabinet or wall mounting – ready to plug-in, easy and quick installation
Safe capturing of escaping vapours and gases at the point where they are released or produced
  • Very low noise, ideal for direct installation at the workplace
  • With connection cable and power cord
  • Connecting fitting DN 75
  • ATEX compliant: C` II 3/-G Ex c llC T4
Specifications
Specifications
CE compliant: C ` II 3/- G Ex c IIC T4
Extraction Unit
Safe capturing of escaping vapours and gases at the point where they are released or produced
200 mm
Powder-coated plain
200 mm
400 mm
400 x 200 x 200 mm
For tall cabinets
HF.EA.8677.
10.1 kg
Extraction Unit