missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Heavy Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
262.00 € - 415.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10654582
|
Lenz Laborglasintrumente
05433137 |
100 mL |
262.00 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
10151471
|
Lenz Laborglasintrumente
05433149 |
250 mL |
343.00 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
10661702
|
Lenz Laborglasintrumente
05433158 |
500 mL |
415.00 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
- For the extraction of liquids with specific heavy solvents
- With insert filter (porosity: 0)
- Made of DURAN™ tubing
Specifications
Fritte P0 |