Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Allihn Condenser
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
254.00 € - 325.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10204631
|
Lenz Laborglasintrumente
05365046 |
200 mL |
325.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
10234581
|
Lenz Laborglasintrumente
05365031 |
70 mL |
271.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
10519581
|
Lenz Laborglasintrumente
05365043 |
150 mL |
322.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
10614012
|
Lenz Laborglasintrumente
05365025 |
30 mL |
254.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
Description
- Extractor in compact and robust design
- With flask connection
- Made of DURAN™ tubing
- Allihn condenser with thread GL 14
- Round bottom flask
Specifications
Duran Glass |