Learn More
Eye Wash Dispenser
Brand:  Safety First E440
Product Code. 12920485
  • 42.05 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The 1997 Approved Code of Practice states that “Where tap water is not readily available for eye irrigation, at least 1L of sterile water or sterile normal saline 0.9% in sealed disposable containers should be provided”.
Specifications
Specifications
Eye Wash Pods
35 cm
Integrated Mirror, Instructions
Wall
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products