Learn More
KARTELL Eye Wash Station
Eye Wash Station is an important safety measure for lab emergencies.
Brand:  KARTELL 0238600
Product Code. 10182661
  • 338.00 € / Pack of 10
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Emergency eye wash station moulded in strong high-impact PS.
  • The directions are clearly printed on the support rack and the bottle can be filled with distilled water or other medicated liquid for ophthalmic use.
  • Eye wash bottle art. 0038300, to be ordered separately, is made of PE provided with an eye cup with snap on cap, carefully designed to allow a quick and immediate flushing to remove foreign matters and prevent serious eye damage.
  • The head has an air-vent valve to assure air refill of free space in the bottle thus preventing draw back of contaminated wash into the bottle.
  • A perforated spray plate in the cup centre breaks up the stream to give the eye a gentle wash.
  • A drain tube releases waste.
Specifications
Specifications
White
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KARTELL Eye Wash Station > Eye Wash Station