Learn More
Fisherbrand™ Eyepiece Pair for Steddy Microscopes
Brand:  Fisherbrand™ 7217.1507
 View more versions of this product
Product Code. 12944361
  • 344.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Eyepiece Pair
  • For STEDDY-T Trinocular Stereo Microscope FB69056
Trinocular configuration allows for the addition of a camera for digital imaging or video camera.
The diamond shaped base provides stability and hand support whilst samples are manipulated on the stage. The zoom ratio of 1x to 4x gives a magnification of 10x to 40x with the standard 10x eyepieces; both with dioptre compensation. Additional eyepieces and corrective lenses allow for a total magnification range from 2.5 to 120x. Both light sources have variable intensity control. The unit comes with frosted glass stage, a blue filter and a heat filter for the incident light source to prevent overheating of the sample.
Robust stereo zoom microscope
  • Integral single piece column stand eliminates vibration and image distortion
  • Ergonomic diamond shaped base
  • Total magnification range 2.5x to 120x
  • Variable intensity transmitted and incident light sources
Specifications
Specifications
Eyepiece
Steddy Microscopes
1 Ea.
13 mm
15X