missing translation for 'onlineSavingsMsg'
Learn More

Plum™ Eyewash Station

Wall station with 500 mL Plum Eye Wash. Particularly suitable for small and mobile workplaces.

Brand:  Plum™ P4611

Product Code. 15791137

  • 58.23 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Visible and readily available first aid
  • Easily mounted on the wall close to the workplace and immediate washing may be needed
  • Ensures quick and efficient eyewash
  • Supplied with pictograms and mirror
  • Use in dust-free and dirt-free areas
Specifications

Specifications

500 mL
300 mm
170 mm
80 mm
500 mL Eyewash
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Plum™ Eyewash Station > Includes: 500 mL Eyewash

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.