Learn More
Gardco™ EZ/3 Series Viscosity Cup #3 with Certificate
Gardco EZ3/C series viscosity cup #3 with certificate is an easy and economical way to measure viscosity.
Brand:  Gardco™ 08700-22
Product Code. 11775359
  • 595.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Supplied with conversion table to determine viscosity in centistokes (cSt)
  • Meets dimensions as specified in ASTM D 4212 standards
  • NIST Traceable certificate
Specifications
Specifications
64 to 596 cSt
Gardco EZ3/C Series Viscosity Cup #3
Cup dimensions as specified in ASTM D 4212 standards; NIST-traceable models conform to MIL STD 5662A, ANSI/NCSL 2540-1; Traceable to NIST-standard oils; NIST-traceable NIST Certificate
46.5 mL