missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ HPDP-Biotin, No-Weigh™ Format
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 1 mg
Unit Size:
50mg
Pack of 10
Description
Thermo Scientific™ EZ-Link HPDP-Biotin is pyridyldithiol-activated, sulfhydryl-reactive biotinylation reagent that conjugates via a cleavable (reversible) disulfide bond to enable use in a variety of purification methods.
Highlights:
- Pyridyldithiol reacts with -SH groups to form a stable disulfide bond
- Absorbance of pyridine-2-thione leaving group at 343nm can be used to determine reaction progress
- Disulfide bond is cleavable using 50mM DTT (Part No. 20291) or 100mM β-Mercaptoethanol (Part No. 35600)
- Extended spacer arm prevents steric hindrance
- Must be dissolved in DMSO or DMF before use
- Biotinylation occurs between pH 6-9
Specifications
Specifications
| Product Type | HPDP-Biotin |
| Quantity | 50 mg |
| Content And Storage | Store at 4°C. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Permeant |
| Chemical Reactivity | Thiol |
| Reactive Moiety | Pyridyldisulfide |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) |
| Label or Dye | Biotin |
| Spacer | Long, Cleavable |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction