missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin, No-Weigh™ Format

Product Code. 10519873 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 2 mg
Unit Size:
50mg
Pack of 10
This item is not returnable. View return policy

Product Code. 10519873

Brand: Thermo Scientific™ 21901BID

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.

Thermo Scientific™ EZ-Link Maleimide-PEG2-Biotin is a mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.

Highlights:

  • Water-soluble; contains polyethylene glycol (PEG) spacer arm
  • Reacts with sulfhydryl (-SH) groups at acidic to neutral pH
  • Transfers hydrophilic PEG spacer arm to final conjugate
  • Forms a thioether bond
TRUSTED_SUSTAINABILITY

Specifications

Product Type Maleimide-PEG2-Biotin
Quantity 50 mg
Content And Storage Store desiccated at 4°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Long, Pegylated
Label Type Biotin & Analogs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.