Learn More
Thermo Scientific™ EZ-Link™ NHS-Biotin
Biotinylate protein amines (e.g., lysines) with a simple, short (13.5A), NHS-ester biotinylation reagent containing a membrane-permeable spacer arm for intracellular labeling.
Brand:  Thermo Scientific™ 20217
Product Code. 10381394
  • 199.00 € / 100mg
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ EZ-Link NHS-Biotin is the simplest NHS-ester activated biotinylation reagent for labeling primary amines (e.g., protein lysines), whose membrane permeability enables it to be used for general intracellular labeling.
Highlights:
  • Forms stable amide bonds
  • Reacts with primary amines at pH 7-9
  • Must be dissolved in DMSO or DMF and can then be transferred to an aqueous reaction solution
  • Contains a shorter spacer arm than NHS-LC-Biotin
  • Penetrates cell membranes because it is uncharged
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NHS-Biotin
Store desiccated at room temperature.
Cell-Permeant
Active Ester, Succinimidyl Ester, NHS Ester
Biotin
Biotin & Analogs
100 mg
EZ-Link™
Amine
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Short