Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG12-Biotin, No-Weigh™ Format
Biotinylate protein amines and maximize solubility of proteins with this very long (56.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.
Brand:  Thermo Scientific™ 21312
 View more versions of this product
Product Code. 11801195
  • 328.00 € / 25mg
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ EZ-Link NHS-PEG12-Biotin is very long, pegylated biotinylation reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Highlights:
  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Excellent purity, molecular weight uniformity and amine-reactivity
  • Long spacer arm reduces steric hindrance when binding to avidin molecules
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NHS-PEG12-Biotin
Store desiccated at -20°C. Shipped at ambient temperature.
Cell-Impermeant
Active Ester, Succinimidyl Ester, NHS Ester
Biotin
Biotin & Analogs
25 mg
EZ-Link™
Amine
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Extra-long, Pegylated