Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
242.00 € - 437.00 €
Specifications
Product Type | Sulfo-NHS-LC-Biotin |
---|---|
Product Line | EZ-Link™ |
Cell Permeability | Cell-Impermeant |
Chemical Reactivity | Amine |
Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906125
|
Thermo Scientific™
A39257 |
10 x 1 mg |
242.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10538723
|
Thermo Scientific™
21335 |
100 mg |
437.00 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
- Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
- Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
- Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
- Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
- Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
- Medium length - spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass
Specifications
Sulfo-NHS-LC-Biotin | |
Cell-Impermeant | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Mid-length |
EZ-Link™ | |
Amine | |
Biotin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.