Learn More
Brady™ EZ Panel Loc Lock Rail
Close panel doors, even when lockouts are in place. The Brady™ EZ Panel Loc System's unique rail system accommodates multiple locks, eliminating the need for hasps during group lockouts.
Brand:  Brady™ 051264
 View more versions of this product
Product Code. 15673487
  • 38.75 € / Pack of 2
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique lock rail system allows panel door to be closed even when lockouts are in place
  • Accommodates multiple locks, eliminating the need for hasps during group lockouts
  • Lock rail design also keeps weight off of the breakers, which protects delicate electrical components from damage
  • Lock rails are made of durable isoplast polymers with acrylic adhesive backing
  • "No shackle" locks with EX Panel Loc keys are sold separately and are especially useful when you want lockout devices to be kept at the point of need
  • A spacing gasket is also available for low profile panels
Specifications
Specifications
Lock Rail
Yellow
4 x 0.8 x 0.7 x in.
2 Pk.
BREAKER BLOCKER RAISED LOCK RAIL 2 PACK
Fiberglass-reinforced Nylon
4 in.