Learn More
Biochrom EZ Read 400 Microplate Reader
EZ Read 400 Microplate Readers with Galapagos for EZ Read Software.
4535.00 €
Specifications
Detector Type | Single Channel Silicon Photodiode |
---|---|
Light Source | Tungsten Halogen Lamp |
Measurement Speed | 25 sec. |
Plate Type | 96-well Plates |
Software Type | Galapagos Software |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
12694795
|
Biochrom
80-4001-40 |
EZ Read 400 ELISA | N/A | ||||||
Description
The EZ Read 400 is a simple and robust microplate reader that is an ideal choice for laboratories which require a workhorse instrument for their routine absorbance assays.
The EZ Read 400 microplate readers are PC-controlled using new, intuitive Galapagos software created by Biochrom to simplify data acquistion and analysis.
EZ Read 400 ELISA and EZ Read 800 Research microplate reader features
- Measures flat, round and v-bottomed 96-well microplates and stripwells with standard ANSI SBS footprints are measured
- Accurate measurements ensured by automatic lamp calibration prior to each reading
- USB 2.0 PC connection
- Interference filters for wavelength selection
- Visible range (400 to 750 nm)
The EZ Read 400 is available in two applictions-focussed models:
- EZ Read 400 ELISA is a dedicated ELISA reader. Comes with 4 filters that can be used to measure most ELISA assays (405, 450, 492 and 620 nm)
- ELISA Assay Substrate
- NPP, ABTS use Filter 405 nm
- OPD use Filter 492 nm
- Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
- Reference wavelength use Filter 620 nm
- ELISA Assay Substrate
- EZ Read 400 Research offers versatility by extending its applications capacity to measuring total protein, cell proliferation assays as well ELISA assays. Comes with 6 filters that can be used to measure ELISA, total protein and cell proliferation assays (405, 450, 492, 562, 570, 595, 620 and 650 nm)
- ELISA Assay Substrate
- NPP, ABTS use Filter 405 nm
- OPD use Filter 492 nm
- Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
- Reference wavelength use Filter 620 nm
- Total Protein Assays
- Bradford use Filter 595 nm
- BCA use Filter 562 nm
- Lowry use Filter 450 nm
- Reference wavelength use Filter 650 nm
- Cell Proliferation/Viability Assays
- MTT use Filter 570 nm
- XTT use Filter 492 nm
- Cell Density use Filter 620 nm
- ELISA Assay Substrate
Galapagos for EZ Read Software's 'click and drag' approach makes usability a key feature:
- Create new plate layouts
- Add multiple assays to a plate
- Add multiple plates per analysis
- Easy to create new methods
- Select only the data you want for data reporting
- Windows 7 & 8 compatibility as well as older Windows OS from XP
- So easy you won't need to export your data into Excel
Specifications
Single Channel Silicon Photodiode | |
25 sec. | |
Galapagos Software | |
0.5% at 1.0 OD (at 450 nm) | |
0.25% and 0.0025 O.D. from 0.1 to 2.5 O.D. (at 492 nm) | |
400 to 750 nm |
Tungsten Halogen Lamp | |
96-well Plates | |
Filters | |
100/240 V, 50/60 Hz, 1.5 A | |
0.25% at 1.0 O.D. (at 450 nm) | |
6.6 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.