missing translation for 'onlineSavingsMsg'
Learn More

Leica Microsystems EZ4 Educational Stereomicroscope (without eyepieces)

High quality, full-featured E-series microscope at an economical price. Leica Microsystems™ EZ4 Educational Stereomicroscope (without eyepieces) features high image quality, color and detail fidelity as well as mechanical precision for exacting control.

Brand:  Leica Microsystems 10447199

Product Code. 12838005

  • 2680.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Educational microscope.
  • Integrated 7-way LED illumination system provides incident, oblique and transmitted light for high quality illumination and contrast.
  • Provides over 20 years of bright LED illumination under normal use.
  • Greenough optical system provides a 3-dimensional view.
Specifications

Specifications

2m Power cable
16X to 70X
3.6 kg
100/240 V 50/60 Hz
LED
1 Each
230 V
4.4:1
225 x 181 x 335 mm
Stereomicroscope
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.