missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EZH2 Monoclonal Antibody (AC22), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ 14-9867-37

 View more versions of this product

Product Code. 17150812

  • 1586.00 € / 2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Description: The monoclonal antibody AC22 recognizes EZH2 (Enhancer of zeste homolog 2), a member of the polycomb group (PcG) proteins responsible for repression of gene expression through recognition and modification of histone methylation and chromatin structure. EZH2 comprises the catalytic subunit of the polycomb repressive complex 2 (PRC2) and directly represses gene expression through methylation of histone H3 on lysine 27 (H3K27) as well as direct protein targets such as GATA4. The PRC2 complex gene repression is necessary for proper development and differentiation. Underexpression of EZH2 is linked to abnormal developmental patterning and a loss of stem cell pluripotency, while overexpression of EZH2 is present in many tumors and correlates with poor prognosis in melanoma, breast, and prostate cancer. Applications Reported: This AC22 antibody has been reported for use in immunoprecipitation, western blotting, and immunocytochemistry. Applications Tested: This AC22 antibody has been tested by immunocytochemistry of methanol-fixed cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EZH2
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q15910, Q61188
EZH2
Affinity chromatography
RUO
14056, 2146, 312299
4°C
Liquid
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
AC22
Unconjugated
EZH2
an; enhancer of zeste 2 polycomb repressive complex 2 subunit; enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; Enx-1; Enx1h; EZH 2; EZH1; EZH2; EZH-2; EZH2b; histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2
Mouse
2 mg
Primary
Human, Mouse, Rat
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.