missing translation for 'onlineSavingsMsg'
Learn More

F-Actin Antibody (NH3), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61610

Product Code. 18669616

  • 348.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

F-Actin Monoclonal antibody specifically detects F-Actin in Human, Mouse, Chicken, Rabbit samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

F-Actin
Monoclonal
1 mg/mL
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
actin, cytoplasmic 2, DFNA26, ACT, ACTG, actin, gamma 1, BRWS2, DFNA20
Human monocytes and the human monocyte-like cell line, U937.
0.1 mL
Developmental Biology
Human, Mouse, Chicken, Rabbit
Purified
Flow Cytometry, ELISA
NH3
Unconjugated
PBS
Mouse
Thiophilic Chromatography
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgM κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
F-Actin Antibody (NH3), Novus Biologicals™ > 0.1 mL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.